Learn More
Biomatik Corporation Mouse Protease Activated Receptor 2 (PAR2) ELISA Kit
Supplier:  Biomatik Corporation EKU06872
Encompass_Preferred
Mouse Protease Activated Receptor 2 (PAR2) ELISA Kit for detecting PAR2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.053ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P55086. Gene ID: 14063. Target Synonyms: F2RL1; F2-RL1; GPR11; PAR2; Coagulation Factor II (thrombin) receptor-Like 1; G-protein coupled receptor 11; Thrombin receptor-like 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8804