Learn More
American Research Products Inc Mouse Protein arginine deimina
Catalog No. 5000326025
Encompass
Mouse Protein-arginine deiminase type-4 (PADI4) ELISA kit from CUSABIO Detection Range 4 7 pg/mL-300 pg/mL Sensitivity 1 17 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-26025 Supplier American Research Products Inc Supplier No. CSBEL017379MO
Mouse Protein-arginine deiminase type-4 (PADI4) ELISA kit from CUSABIO Detection Range 4 7 pg/mL-300 pg/mL Sensitivity 1 17 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich