Learn More
American Research Products Inc Mouse Protein argonaute 2 EIF
Catalog No. 5000327949
Encompass
Mouse Protein argonaute-2 (EIF2C2) ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27949 Supplier American Research Products Inc Supplier No. CSBEL007520MO
Mouse Protein argonaute-2 (EIF2C2) ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich