Learn More
American Research Products Inc Mouse Protein CYR61 CYR61 EL
Catalog No. 5000327944
Encompass
Mouse Protein CYR61 (CYR61) ELISA kit from CUSABIO Detection Range 18 75 pg/mL-1200 pg/mL Sensitivity 4 68 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27944 Supplier American Research Products Inc Supplier No. CSBEL006463MO
Mouse Protein CYR61 (CYR61) ELISA kit from CUSABIO Detection Range 18 75 pg/mL-1200 pg/mL Sensitivity 4 68 pg/mL Sample serum plasma tissue homogenates Method Sandwich