Learn More
American Research Products Inc Mouse Protein delta homolog 1
Catalog No. 5000328238
Encompass
Mouse Protein delta homolog 1 (DLK1) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28238 Supplier American Research Products Inc Supplier No. CSBEL006945MO
Mouse Protein delta homolog 1 (DLK1) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich