Learn More
Biomatik Corporation Mouse Protein Disulfide Isomerase (PDI) ELISA Kit
Supplier:  Biomatik Corporation EKU06901
Encompass_Preferred
Mouse Protein Disulfide Isomerase (PDI) ELISA Kit for detecting PDI in tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.064ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: D3Z6P0. Gene ID: 69191. Target Synonyms: PDI-A2; PDA2, PDIA2, PDIR; PDIP; Protein Disulfide Isomerase,Pancreatic; Protein Disulfide Isomerase-Associated 2; Pancreas-specific protein disulfide isomerase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5626