Learn More
American Research Products Inc Mouse Protein disulfide isomer
Supplier:  American Research Products Inc CSBEL017722MO
Encompass
Mouse Protein disulfide-isomerase A4 (PDIA4) ELISA kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27386