Learn More
American Research Products Inc Mouse Protein DJ 1 PARK7 ELI
Catalog No. 5000327999
Encompass
Mouse Protein DJ-1 (PARK7) ELISA kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27999 Supplier American Research Products Inc Supplier No. CSBEL017454MO
Mouse Protein DJ-1 (PARK7) ELISA kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich