Learn More
Biomatik Corporation Mouse Protein Interacting With C Kinase 1 (PICK1) ELISA Kit
Supplier:  Biomatik Corporation EKU06908
Encompass_Preferred
Mouse Protein Interacting With C Kinase 1 (PICK1) ELISA Kit for detecting PICK1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.053ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q62083. Target Synonyms: PRKCABP; Protein kinase C,Alpha Binding Protein; Protein Interacting With Protein Kinase C Alpha 1; Protein kinase C-alpha-binding protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5084