Learn More
Biomatik Corporation Mouse Protein Kinase, AMP Activated Alpha 1 (AMPK Alpha 1) ELISA Kit
Supplier:  Biomatik Corporation EKU08468
Encompass_Preferred
Mouse Protein Kinase, AMP Activated Alpha 1 (AMPK Alpha 1) ELISA Kit for detecting AMPK Alpha 1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.57ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q5EG47. Gene ID: 105787. Target Synonyms: PRKAa1; AMPK; AMPKa1; PKA; ACACA kinase; 5'-AMP-Activated Protein Kinase Catalytic Subunit Alpha-1; Acetyl-CoA carboxylase kinase; Hydroxymethylglutaryl-CoA reductase kinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4670