Learn More
American Research Products Inc Mouse Protein notum homolog N
Catalog No. 5000328119
Encompass
Mouse Protein notum homolog (NOTUM) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates cell culture supernates Method Sandwich
Catalog No. 50-003-28119 Supplier American Research Products Inc Supplier No. CSBEL015955MO
Mouse Protein notum homolog (NOTUM) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates cell culture supernates Method Sandwich