Learn More
American Research Products Inc Mouse Protein S100 A4 S100A4
Catalog No. 5000327153
Encompass
Mouse Protein S100-A4 (S100A4) ELISA kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-27153 Supplier American Research Products Inc Supplier No. CSBEL020632MO
Mouse Protein S100-A4 (S100A4) ELISA kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich