Learn More
Biomatik Corporation Mouse Protein Tyrosine Phosphatase Receptor Type C (CD45) ELISA Kit
Supplier:  Biomatik Corporation EKU11689
Encompass_Preferred
Mouse Protein Tyrosine Phosphatase Receptor Type C (CD45) ELISA Kit for detecting CD45 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.127ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P06800. Gene ID: 19264. Target Synonyms: PTPRC; PTPR-C; LCA; B220; GP180; LY5; T200; Leukocyte common antigen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7718