Learn More
American Research Products Inc Mouse Protein Wnt 16 WNT16 E
Catalog No. 5000328285
Encompass
Mouse Protein Wnt-16 (WNT16) ELISA kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-28285 Supplier American Research Products Inc Supplier No. CSBEL026132MO
Mouse Protein Wnt-16 (WNT16) ELISA kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich