Learn More
American Research Products Inc Mouse Protein Wnt 2 WNT2 ELI
Catalog No. 5000327247
Encompass
Mouse Protein Wnt-2 (WNT2) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27247 Supplier American Research Products Inc Supplier No. CSBEL026133MO
Mouse Protein Wnt-2 (WNT2) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich