Learn More
American Research Products Inc Mouse Protein Wnt 5a WNT5A E
Catalog No. 5000327351
Encompass
Mouse Protein Wnt-5a (WNT5A) ELISA kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-27351 Supplier American Research Products Inc Supplier No. CSBEL026138MO
Mouse Protein Wnt-5a (WNT5A) ELISA kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich