Learn More
American Research Products Inc Mouse Protein Wnt 7b WNT7B E
Catalog No. 5000327401
Encompass
Mouse Protein Wnt-7b (WNT7B) ELISA kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27401 Supplier American Research Products Inc Supplier No. CSBEL026142MO
Mouse Protein Wnt-7b (WNT7B) ELISA kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich