Learn More
Biomatik Corporation Mouse Protein Z (PROZ) ELISA Kit
Supplier:  Biomatik Corporation EKU06953
Encompass_Preferred
Mouse Protein Z (PROZ) ELISA Kit for detecting PROZ in plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.067ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9CQW3. Gene ID: 66901. Target Synonyms: PRO-Z; PZ; Vitamin K Dependent Protein Z; VKDPZ; VKDP-Z. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4482