Learn More
American Research Products Inc Mouse Proto oncogene Wnt 1 WN
Supplier:  American Research Products Inc CSBEL026128MO
Encompass
Mouse Proto-oncogene Wnt-1 (WNT1) ELISA kit from CUSABIO Detection Range 18 75 pg/mL-1200 pg/mL Sensitivity 4 68 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27254