Learn More
American Research Products Inc Mouse Proto oncogene Wnt 3 WN
Catalog No. 5000327941
Encompass
Mouse Proto-oncogene Wnt-3 (WNT3) ELISA kit from CUSABIO Detection Range 39 pg/ml-2500 pg/ml Sensitivity 9 75 pg/ml Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27941 Supplier American Research Products Inc Supplier No. CSBEL026135MO
Mouse Proto-oncogene Wnt-3 (WNT3) ELISA kit from CUSABIO Detection Range 39 pg/ml-2500 pg/ml Sensitivity 9 75 pg/ml Sample serum plasma tissue homogenates cell lysates Method Sandwich