Learn More
Biomatik Corporation Mouse Protocadherin 15 (PCDH15) ELISA Kit
Supplier:  Biomatik Corporation EKU06970
Encompass_Preferred
Mouse Protocadherin 15 (PCDH15) ELISA Kit for detecting PCDH15 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.065ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q99PJ1. Gene ID: 11994. Target Synonyms: USH1F; DFNB23; CDHR15; Deafness,Autosomal Recessive 23; Cadherin-Related Family Member 15. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6106