Learn More
Cayman Chemical Mouse PRoxISME ProlIfRatr
SDP
Catalog No. 502972684
Encompass_Preferred
mPPARα Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. M00111-32, M00111]
Catalog No. 50-297-2684 Supplier Cayman Chemical Supplier No. 157623X32 WELL
mPPARα Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. M00111-32, M00111]