Learn More
Sino Biological Anti-PSAP/Prosaposin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 51192RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse PSAP (rM PSAP; Catalog#51192-M08H; NP_035309.3; Met1-Asn557). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1769