Learn More
Sino Biological Anti-PSAP/Prosaposin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 51192T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse PSAP/Prosaposin (rM PSAP/Prosaposin; Catalog#51192-M08H; NP_035309.3; Met1-Asn557). PSAP/Prosaposin specific IgG was purified by Mouse PSAP/Prosaposin affinity chromatography.
Catalog No. 50-241-0104