Learn More
Sino Biological Mouse PSCA Protein 1MG (Sino Biological)
Supplier:  Sino Biological 5A1731M08H1
Encompass_Preferred
A DNA sequence encoding the Mouse PSCA (P57096) (Met1-Asn95) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-3659