Learn More
Sino Biological Mouse PSCA Protein 20ug (Sino Biological)
Supplier:  Sino Biological 5A1731M49HB20
Encompass_Preferred
A DNA sequence encoding the Mouse PSCA (P57096) (Met1-Asn95) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-237-2850