Learn More
Biomatik Corporation Mouse P-Selectin Glycoprotein Ligand 1 (PSGL1) ELISA Kit
Supplier:  Biomatik Corporation EKU06975
Encompass_Preferred
Mouse P-Selectin Glycoprotein Ligand 1 (PSGL1) ELISA Kit for detecting PSGL1 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.066ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q62170. Target Synonyms: CD162; SELPLG; CLA; Selectin P Ligand. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7456