Learn More
Biomatik Corporation Mouse P-Selectin (SELP) CLIA Kit
Supplier:  Biomatik Corporation EKU08900
Encompass_Preferred
Mouse P-Selectin (SELP) CLIA Kit for detecting SELP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 13.7-10,000pg/ml. Sensitivity: 5.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q01102. Gene ID: 20344. Target Synonyms: CD62P; Selectin, Platelet; PSEL; GMP140; LECAM3; GRMP; GMRP; PADGEM; Leukocyte-endothelial cell adhesion molecule 3; Platelet activation dependent granule-external membrane. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8599