Learn More
Sino Biological Mouse PSMA / FOLH1 Protein (His Tag) 100ug
Supplier:  Sino Biological 50214M07H100
Encompass_Preferred
A DNA sequence encoding the mouse FOLH1 (NP_058050.3) extracellular domain (Ile 44-Ala 752) was expressed, fused with a polyhistidine tag at the N-terminus.
Catalog No. 50-224-7064