Learn More
Sino Biological Anti-Brk Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50710T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse PTK6 / Brk (Catalog#50710-M20B; Q64434; Met1-Val451). PTK6 / Brk specific IgG was purified by Mouse PTK6 / Brk affinity chromatography.
Catalog No. 50-255-3129