Learn More
ACROBiosystems ACROBiosystems Mouse PTK7 / CCK4 Protein, His Tag
Supplier:  ACROBiosystems PT7M52H31MG
Encompass_Preferred
Recombinant Protein;1MG;Mouse PTK7, His Tag (PT7-M52H3) is expressed from human 293 cells (HEK293). It contains AA Ala 23 - Thr 696 (Accession # Q8BKG3-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 76.5 kDa.
Catalog No. 50-210-6379