Learn More
Sino Biological Recombinant Anti-TC-PTP/PTPN2 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50800R001100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse PTPN2 / TC-PTP (rM PTPN2 / TC-PTP; Catalog#50800-M07B; Q06180-1; Ser2-Asn314).
Catalog No. 50-255-3078