Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Pure-Blot anti-Rabbit IgG (H+L) Secondary Antibody (eB182), DyLight 800, Novus Biologicals™
SDP

Catalog No. NB120521
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB120521 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB120521 Supplier R&D Systems Supplier No. NBP311666

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

IgG Monoclonal antibody specifically detects IgG in Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, ELISA

Specifications

Antigen IgG
Applications Dot Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone eB182
Concentration LYOPH
Conjugate DyLight 800
Dilution Western Blot 1:1000, Flow Cytometry 1:2000 - 1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:500 - 1:2500, Immunoprecipitation, Fluorophore-linked immunosorbent assay
Formulation Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/ml Polyethylene Glycol (PEG-8000)
Gene Alias IP Detection Reagent
Host Species Mouse
Immunogen Rabbit IgG
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Epitope Tags, Immunology
Primary or Secondary Secondary
Gene ID (Entrez) 3500
Reconstitution Reconstitute with 100 ul deionized water (or equivalent).
Target Species Rabbit
Content And Storage Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.