Learn More
Sino Biological Recombinant Mouse CD155/PVR Protein (ECD, hFc & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 50259M41HB20
Encompass_Preferred
A DNA sequence encoding the mouse PVR (NP_081790.1) (Met1-Arg345) was expressed with a C-terminal Fc region of human IgG1 tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-3321