Learn More
ACROBiosystems ACROBiosystems Mouse PVRIG Protein, Mouse IgG2a Fc Tag
Supplier:  ACROBiosystems PVGM5253100UG
Encompass_Preferred
Recombinant Protein;100UG;Mouse PVRIG, Mouse IgG2a Fc Tag (PVG-M5253) is expressed from human 293 cells (HEK293). It contains AA Ser 35 - Asp 165 (Accession # A0A1B0GS01-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 41.0 kDa. The protein migrates as 50-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6237