Learn More
Biomatik Corporation Mouse Pyruvate Dehydrogenase Beta (PDHb) ELISA Kit
Supplier:  Biomatik Corporation EKU06988
Encompass_Preferred
Mouse Pyruvate Dehydrogenase Beta (PDHb) ELISA Kit for detecting PDHb in tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.066ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9D051. Gene ID: 68263. Target Synonyms: PDH-B; PHE1B; PHE1-B; Pyruvate Dehydrogenase (lipoamide) Beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5464