Learn More
Biomatik Corporation Mouse Pyruvate Dehydrogenase Kinase 4 (PDK4) ELISA Kit
Supplier:  Biomatik Corporation EKU06990
Encompass_Preferred
Mouse Pyruvate Dehydrogenase Kinase 4 (PDK4) ELISA Kit for detecting PDK4 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.127ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O70571. Gene ID: 27273. Target Synonyms: [Pyruvate dehydrogenase (acetyl-transferring) ] kinase isozyme 4, mitochondrial; Pyruvate Dehydrogenase Kinase Isozyme 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5282