Learn More
Biomatik Corporation Mouse Pyruvate Dehydrogenase Kinase Isozyme 3 (PDK3) ELISA Kit
Supplier:  Biomatik Corporation EKU12426
Encompass_Preferred
Mouse Pyruvate Dehydrogenase Kinase Isozyme 3 (PDK3) ELISA Kit for detecting PDK3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q922H2. Gene ID: 236900. Target Synonyms: PDHK3; [Pyruvate dehydrogenase (acetyl-transferring) ] kinase isozyme 3, mitochondrial. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4894