Learn More
Biomatik Corporation Mouse Pyruvate kinase isozymes R/L (PKLR) ELISA Kit
Supplier:  Biomatik Corporation EKU11383
Encompass_Preferred
Mouse Pyruvate kinase isozymes R/L (PKLR) ELISA Kit for detecting PKLR in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.62ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P53657. Target Synonyms: PK; PyK; PK1; PKL; Pyruvate Kinase Isozymes R/L; Glutamic--pyruvic transaminase 1; R-type/L-type pyruvate kinase; Red cell/liver pyruvate kinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6258