Learn More
Abcam Mouse and Rabbit Specific HRP/DAB IHC Detection Kit - Micro-polymer, 15ML
Catalog No. NC1893798
Encompass_Preferred
Rabbit/mouse dual, HRP/DAB detection IHC kit is a colorimetric micro-polymer assay kit used to detect mouse and rabbit primary antibodies in immunohistochemistry (IHC). - Cited in over 65 publications - No need for endogenous biotin blocking - Includes ready-to-use blocking reagents - Higher sensitivity than streptavidin-biotin methods
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1893798 Supplier Abcam Supplier No. AB23646615ML
Rabbit/mouse dual, HRP/DAB detection IHC kit is a colorimetric micro-polymer assay kit used to detect mouse and rabbit primary antibodies in immunohistochemistry (IHC). - Cited in over 65 publications - No need for endogenous biotin blocking - Includes ready-to-use blocking reagents - Higher sensitivity than streptavidin-biotin methods
The product is subject to the following: Abcam Restricted Use Statement