Learn More
Biomatik Corporation Mouse RAB5A, Member RAS Oncogene Family (RAB5A) CLIA Kit
Supplier:  Biomatik Corporation EKU10024
Encompass_Preferred
Mouse RAB5A, Member RAS Oncogene Family (RAB5A) CLIA Kit for detecting RAB5A in tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 13.7-10,000pg/ml. Sensitivity: 6.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9CQD1. Gene ID: 271457. Target Synonyms: RAB5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7156