Learn More
Sino Biological Anti-RAB9A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 106498T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Mouse RAB9A, and purified by antigen affinity chromatography.
Catalog No. 50-240-9150