Learn More
Diagnostic Biosystems MOUSE/RABBIT AP DETECTION REAG UnoVue™ Mouse/Rabbit AP Detection Reagent
Catalog No. NC2027032
Encompass_Preferred
The UnoVue Mouse/Rabbit AP Detection reagent is suitable for use with all mouse and rabbit antibodies, both monoclonal and polyclonal. The reagents can be used for manual staining or with automated staining instruments and are well suited for multiplex immunohistochemical staining assays.
Catalog No. NC2027032 Supplier Diagnostic Biosystems Supplier No. MRUAP100
The UnoVue Mouse/Rabbit AP Detection reagent is suitable for use with all mouse and rabbit antibodies, both monoclonal and polyclonal. The reagents can be used for manual staining or with automated staining instruments and are well suited for multiplex immunohistochemical staining assays.