Learn More
FUJIFILM BIOSCIENCES INC Mouse RANK-Ligand , Recombinant Protein, Shenandoah Biotechnology, 1 mg vial
Supplier:  FUJIFILM BIOSCIENCES INC 200041MG
Encompass_Preferred
Receptor activator of nuclear factor kappa-B Ligand (RANK Ligand) is a cell-bound marker related to the tumor necrosis factor (TNF) family of proteins. RANK Ligand plays a critical role in bone metabolism and osteoclast differentiation. T cell expression of RANK Ligand promotes dendritic cell maturation.
Catalog No. 50-990-803
Edge
Add to Cart
Request a Quote