Learn More
Bioworld MOUSE RANTES (CCL5) (RECOM
Supplier:  Bioworld 220605801
Encompass_Preferred
MOUSE RANTES (CCL5), (RECOMBINANT): Recombinant Mouse Rantes (CCL5) Source: E.Coli Description: Recombinant Mouse Rantes produced in E.Coli is a single, non-glycosylated polypeptide chain containing 68 amino acids and having a molecu...
Catalog No. 50-253-680