Learn More
Cayman Chemical Mouse RAR-related Orphan Receptor, Gamma Reporter Assay System, 96 Well
SDP
Catalog No. 502994081
Encompass_Preferred
mRORγ Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments.[INDGO Catalog Nos. M04001-32, M04001]
Catalog No. 50-299-4081 Supplier Cayman Chemical Supplier No. 2140196 WELL
mRORγ Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments.[INDGO Catalog Nos. M04001-32, M04001]