Learn More
Biomatik Corporation Mouse Ras Homolog Gene Family, Member A (RHOA) ELISA Kit
Supplier:  Biomatik Corporation EKU12226
Encompass_Preferred
Mouse Ras Homolog Gene Family, Member A (RHOA) ELISA Kit for detecting RHOA in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9QUI0. Gene ID: 11848. Target Synonyms: ARH12; ARHA; RHO12; RHOH12; Rho cDNA clone 12. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7999