Learn More
Alpco Ltd Mouse / Rat Acylated Ghrelin ELISA
SDP
Supplier:  Alpco Ltd 325117
Encompass_Preferred
The Mouse / Rat Acylated Ghrelin ELISA is for the quantitative determination of acylated ghrelin in mouse and rat plasma. For Research Use only. Not For In Vitro Diagnostic Use.
Dynamic range
1.95-250 pg/mL
Sensitivity
0.7 pg/mL
Sample type
Plasma
Catalog No. NC2332905