Learn More
Calbiotech Inc Mouse/Rat Cotinine ELISA kit, 96 Tests (12x8 breakable strip wells), Standard Range: 5-100 ng/mL, Analytical Sensitivity: 1 ng/mL, Sample Volume: 10 μL/well, Storage Temperature: 2-8 °C
Supplier:  Calbiotech Inc CO096D100
Encompass_Preferred
Mouse/Rat Cotinine ELISA kit, 96 Tests (12x8 breakable strip wells), Standard Range: 5-100 ng/mL, Analytical Sensitivity: 1 ng/mL, Sample Volume: 10 μL/well, Storage Temperature: 2-8 °C
Catalog No. NC1093812