Learn More
Alpco Ltd Mouse / Rat Growth Hormone ELISA
SDP
Supplier:  Alpco Ltd 22GHOMSE01
Encompass
Mouse / Rat Growth Hormone ELISA is intended to be used for the measurement of Growth Hormone in mouse and rat serum and plasma samples for research use.
  • Includes standards and controls
  • Requires only 3 hours of incubation time
  • Small sample size: 10 µL of sample per well
For research use only. Not for use in diagnostic procedures.
Catalog No. 50-442-151